siRNA Details
Gene Symbol: Ddr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166022.1 | NP_001159494.1 | 8 | 1172 | ||
NM_013137.2 | NP_037269.2 | 8 | 1259 | |||
GenBank mRNA | BC166604.1 | - | 8 | 1259 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
