siRNA Details
Gene Symbol: Plat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013151.2 | NP_037283.2 | 11 | 1282 | ||
XM_006253347.2 | XP_006253409.1 | 11 | 1354 | |||
XM_008771348.1 | XP_008769570.1 | 12 | 1412 | |||
GenBank mRNA | BC061565.1 | - | 11 | 1282 | 6920438 | |
M23697.1 | - | 10 | 1206 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
