siRNA Details
Gene Symbol: Slc18a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013152.2 | NP_037284.2 | 6 | 882 | ||
XM_006252993.2 | XP_006253055.1 | 5 | 1082 | |||
XM_008771066.1 | XP_008769288.1 | 3 | 668 | |||
XM_008771067.1 | XP_008769289.1 | 5 | 1082 | |||
GenBank mRNA | AY168444.1 | - | 3 | 135 | ||
M97380.1 | - | 6 | 911 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
