siRNA Details
Gene Symbol: Vldlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013155.2 | NP_037287.2 | 11 | 1718 | ||
XM_006231197.2 | XP_006231259.1 | 12 | 2023 | |||
XM_006231198.2 | XP_006231260.1 | 12 | 1752 | |||
XM_006231199.2 | XP_006231261.1 | 11 | 1626 | |||
GenBank mRNA | L35767.1 | - | 11 | 1718 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013703, NM_001161420, AK076172, L33417, BC013622, and others.
Gene Ontology Categories:
