siRNA Details
Gene Symbol: Mxi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013160.2 | NP_037292.2 | 5 | 455 | ||
XM_006231604.1 | XP_006231666.1 | 5 | 1014 | |||
XM_006231605.1 | XP_006231667.1 | 5 | 426 | |||
XM_006231606.1 | XP_006231668.1 | 5 | 585 | |||
GenBank mRNA | AF003008.1 | - | 5 | 408 | ||
BC081824.1 | - | 5 | 455 | 7133853 |

More Information
Other Species Targeted

Gene Ontology Categories:
