siRNA Details
Gene Symbol: Gucy2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013170.1 | NP_037302.1 | 1 | 141 | ||
GenBank mRNA | M55636.1 | - | 1 | 141 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
