siRNA Details
Gene Symbol: Syt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033680.2 | NP_001028852.2 | 5, 6 | 698 | ||
XM_008765352.1 | XP_008763574.1 | 2, 3 | 680 | |||
GenBank mRNA | AJ617615.1 | - | 5, 6 | 698 | ||
DQ181550.2 | - | 2, 3 | 366 | |||
X52772.1 | - | 5, 6 | 881 |

More Information
Other Species Targeted

Gene Ontology Categories:
