siRNA Details
Gene Symbol: Tgfb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013174.2 | NP_037306.1 | 6 | 1408 | ||
XM_006240366.1 | XP_006240428.1 | 7 | 1752 | |||
GenBank mRNA | BC092195.1 | - | 6 | 1395 | 7313608 | |
U03491.1 | - | 6 | 1408 |

More Information
Other Species Targeted

Gene Ontology Categories:
