siRNA Details
Gene Symbol: Scg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013175.3 | NP_037307.1 | 5, 6 | 579 | ||
XM_006234701.2 | XP_006234763.1 | 5, 6 | 652 | |||
GenBank mRNA | BC061560.1 | - | 5, 6 | 583 | 6918342 | |
FQ212495.1 | - | 5, 6 | 579 | |||
FQ213081.1 | - | 5, 6 | 605 | |||
FQ213100.1 | - | 5, 6 | 605 | |||
M63901.1 | - | 5, 6 | 577 |

More Information
Other Species Targeted

Gene Ontology Categories:
