siRNA Details
Gene Symbol: Acp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270889.1 | NP_001257818.1 | 5 | 1012 | ||
NM_019144.2 | NP_062017.2 | 5 | 954 | |||
XM_006242691.2 | XP_006242753.1 | 7 | 1660 | |||
XM_006242692.2 | XP_006242754.1 | 6 | 1605 | |||
XM_006242693.2 | XP_006242755.1 | 5 | 1471 | |||
XM_006242694.2 | XP_006242756.1 | 5 | 908 | |||
GenBank mRNA | BC078847.1 | - | 5 | 1012 | 7110955 | |
M76110.1 | - | 5 | 952 |
