siRNA Details
Gene Symbol: Pygb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013188.1 | NP_037320.1 | 15 | 1896 | ||
GenBank mRNA | BC166475.1 | - | 1852 | 9100353 | ||
CK357665.1 | - | 4 | 388 | |||
L10668.1 | - | 15 | 1796 | |||
M27726.1 | - | 2 | 80 |
