siRNA Details
Gene Symbol: Nf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013193.1 | NP_037325.1 | 14 | 1894 | ||
XM_006251188.2 | XP_006251250.1 | 14 | 1944 | |||
GenBank mRNA | BP499772.1 | - | 4 | 471 | ||
U17266.1 | - | 1 | 25 | |||
U61772.1 | - | 14 | 1488 |
