siRNA Details
Gene Symbol: Clcn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017106.1 | NP_058802.1 | 3 | 342 | ||
GenBank mRNA | D50497.1 | - | 3 | 342 | ||
Z56277.1 | - | 3 | 294 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
