siRNA Details
Gene Symbol: Cd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175577.3 | NP_783167.2 | 11 | 1670 | ||
XM_006231021.2 | XP_006231083.1 | 11 | 1777 | |||
XM_008760218.1 | XP_008758440.1 | 10 | 1660 | |||
XM_008760219.1 | XP_008758441.1 | 10 | 1473 | |||
GenBank mRNA | AF488727.1 | - | 11 | 1609 | ||
BC089767.1 | - | 10 | 1553 | 7383438 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
