siRNA Details
Gene Symbol: Ogt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017107.2 | NP_058803.2 | 8 | 946 | ||
XM_006257056.1 | XP_006257118.1 | 7 | 1081 | |||
XM_006257057.2 | XP_006257119.1 | 8 | 1023 | |||
XM_006257058.1 | XP_006257120.1 | 7 | 1051 |
