siRNA Details
Gene Symbol: Tfec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022379.1 | NP_071774.1 | 3 | 484 | ||
XM_006236128.2 | XP_006236190.1 | 2 | 750 | |||
GenBank mRNA | L08812.1 | - | 3 | 484 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tfec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022379.1 | NP_071774.1 | 3 | 484 | ||
XM_006236128.2 | XP_006236190.1 | 2 | 750 | |||
GenBank mRNA | L08812.1 | - | 3 | 484 |