siRNA Details
Gene Symbol: Acot7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146061.1 | NP_001139533.1 | 7 | 887 | ||
NM_013214.4 | NP_037346.2 | 7 | 815 | |||
XM_006239447.2 | XP_006239509.1 | 7 | 915 | |||
XM_006239448.1 | XP_006239510.1 | 7 | 823 | |||
GenBank mRNA | BC087716.1 | - | 7 | 767 | 7316866 | |
FQ212652.1 | - | 4 | 366 | |||
FQ231740.1 | - | 4 | 366 | |||
Y09332.1 | - | 7 | 715 |
