siRNA Details
Gene Symbol: Acot7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146061.1 | NP_001139533.1 | 4 | 518 | ||
NM_013214.4 | NP_037346.2 | 4 | 446 | |||
XM_006239447.2 | XP_006239509.1 | 4 | 546 | |||
XM_006239448.1 | XP_006239510.1 | 4 | 454 | |||
GenBank mRNA | BC087716.1 | - | 4 | 398 | 7316866 | |
D88891.1 | - | 4 | 518 | |||
U49694.1 | - | 4 | 426 | |||
Y09332.1 | - | 4 | 346 |

More Information
Other Species Targeted

Gene Ontology Categories:
