siRNA Details
Gene Symbol: Bsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019146.2 | NP_062019.2 | 5 | 3930 | ||
GenBank mRNA | Y16563.1 | - | 5 | 3933 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
