siRNA Details
Gene Symbol: Snn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034083.1 | NP_001029255.1 | 2 | 984 | ||
GenBank mRNA | BC100111.1 | - | 2 | 984 | 7380565 | |
M81639.1 | - | 2 | 967 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
