siRNA Details
Gene Symbol: Grn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145842.1 | NP_001139314.1 | 8 | 815 | ||
NM_017113.2 | NP_058809.2 | 8 | 815 | |||
XM_008767981.1 | XP_008766203.1 | 8 | 896 | |||
GenBank mRNA | BC072469.1 | - | 8 | 809 | 7098196 | |
M97750.1 | - | 7 | 770 | |||
X62322.1 | - | 8 | 789 |

More Information
Other Species Targeted

Gene Ontology Categories:
