siRNA Details
Gene Symbol: Myog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017115.2 | NP_058811.2 | 2, 3 | 584 | ||
GenBank mRNA | AF054894.1 | - | 2, 3 | 550 | ||
M24393.1 | - | 2, 3 | 583 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
