siRNA Details
Gene Symbol: Amelx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271073.1 | NP_001258002.1 | 3 | 149 | ||
NM_001271074.1 | NP_001258003.1 | 3 | 149 | |||
NM_001271075.1 | NP_001258004.1 | 3 | 149 | |||
NM_001271076.1 | NP_001258005.1 | 3 | 149 | |||
NM_001271077.1 | NP_001258006.1 | 3 | 149 | |||
NM_001271078.1 | NP_001258007.1 | 3 | 149 | |||
NM_019154.1 | NP_062027.1 | 2 | 98 | |||
GenBank mRNA | U01245.2 | - | 2 | 108 | ||
U07054.1 | - | 2 | 90 | |||
U51195.1 | - | 3 | 150 | |||
U60562.1 | - | 3 | 150 | |||
U60563.1 | - | 1 | 30 | |||
U60564.1 | - | 3 | 145 | |||
U67130.1 | - | 2 | 98 |

More Information
Other Species Targeted

Gene Ontology Categories:
