siRNA Details
Gene Symbol: Vtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019156.2 | NP_062029.2 | 7 | 1335 | ||
GenBank mRNA | AY318963.1 | - | 1332 | |||
BC105821.1 | - | 7 | 1335 | 7371104 | ||
FQ209609.1 | - | 7 | 1348 | |||
FQ209682.1 | - | 7 | 1343 | |||
FQ209762.1 | - | 3 | 509 | |||
FQ210146.1 | - | 5 | 955 | |||
FQ210518.1 | - | 7 | 1340 | |||
FQ211325.1 | - | 3 | 469 | |||
FQ218318.1 | - | 7 | 1347 | |||
FQ218401.1 | - | 5 | 907 | |||
FQ219297.1 | - | 7 | 1349 | |||
U44845.1 | - | 7 | 1325 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
