siRNA Details
Gene Symbol: Hfe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173434.1 | NP_001166905.1 | 4 | 764 | ||
NM_001173435.1 | NP_001166906.1 | 3 | 464 | |||
NM_053301.4 | NP_445753.1 | 5 | 1028 | |||
GenBank mRNA | AF176534.1 | - | 4 | 684 | ||
AF176535.1 | - | 3 | 384 | |||
AJ001517.1 | - | 5 | 1067 | |||
FQ219597.1 | - | 4 | 778 |
