siRNA Details
Gene Symbol: Inhba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017128.2 | NP_058824.1 | 3 | 1384 | ||
XM_006254001.2 | XP_006254063.1 | 4 | 5043 | |||
XM_008771712.1 | XP_008769934.1 | 3 | 4961 | |||
XM_008771713.1 | XP_008769935.1 | 5 | 6751 | |||
XM_008771714.1 | XP_008769936.1 | 5 | 3531 | |||
XM_008771715.1 | XP_008769937.1 | 5 | 3433 | |||
XM_008771716.1 | XP_008769938.1 | 5 | 1811 | |||
GenBank mRNA | M37482.1 | - | 3 | 1376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
