siRNA Details
Gene Symbol: Syngr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019166.2 | NP_062039.1 | 3 | 467 | ||
XM_006241971.2 | XP_006242033.1 | 3 | 472 | |||
GenBank mRNA | DN932005.1 | - | 3 | 429 | ||
DV724604.1 | - | 2 | 273 | |||
U39549.1 | - | 3 | 436 |

More Information
Other Species Targeted

Gene Ontology Categories:
