siRNA Details
Gene Symbol: Ptma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021740.1 | NP_068508.1 | 5 | 1130 | ||
GenBank mRNA | BC061972.1 | - | 5 | 1142 | 5623463 | |
BC092569.1 | - | 5 | 1154 | 7307282 | ||
FQ210626.1 | - | 5 | 1163 | |||
FQ212611.1 | - | 5 | 1164 | |||
FQ214221.1 | - | 5 | 1158 | |||
FQ215633.1 | - | 5 | 1163 | |||
FQ219686.1 | - | 5 | 1164 | |||
FQ220677.1 | - | 5 | 1163 | |||
FQ221801.1 | - | 5 | 1163 | |||
FQ225103.1 | - | 5 | 1167 | |||
FQ226946.1 | - | 5 | 1163 | |||
FQ227835.1 | - | 5 | 1165 | |||
FQ231794.1 | - | 5 | 1169 | |||
FQ232039.1 | - | 5 | 1169 | |||
M86564.1 | - | 5 | 1130 | |||
X55326.1 | - | 5 | 1130 |
Gene Symbol: RGD1560723 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763685.1 | XP_008761907.1 | 1 | 2745 |

More Information
Other Species Targeted

Gene Ontology Categories:
