siRNA Details
Gene Symbol: Cbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019170.2 | NP_062043.1 | 450 | |||
GenBank mRNA | BC105893.1 | - | 502 | 7366019 | ||
D89069.1 | - | 443 | ||||
X84349.1 | - | 450 |
Gene Symbol: LOC100360507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751016.3 | XP_003751064.2 | 2, 3 | 560 |
Gene Symbol: LOC102556347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248109.2 | XP_006248171.1 | 560 |
