siRNA Details
Gene Symbol: Nfib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031566.1 | NP_113754.1 | 5 | 751 | ||
XM_006238335.2 | XP_006238397.1 | 5 | 1416 | |||
XM_006238336.2 | XP_006238398.1 | 5 | 1416 | |||
XM_006238339.2 | XP_006238401.1 | 5 | 1414 | |||
XM_008763762.1 | XP_008761984.1 | 5 | 1158 | |||
GenBank mRNA | AB012230.2 | - | 5 | 751 | ||
AB012231.2 | - | 5 | 751 | |||
AB012232.2 | - | 5 | 751 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001113209, NM_001286131, NM_001113210, NM_008687, NM_001286127, and others.
Gene Ontology Categories:
