siRNA Details
Gene Symbol: Penk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017139.1 | NP_058835.1 | 2 | 568 | ||
XM_006237835.1 | XP_006237897.1 | 3 | 421 | |||
GenBank mRNA | BC083563.1 | - | 2 | 559 | 7112833 | |
FQ213928.1 | - | 3 | 401 | |||
M28263.1 | - | 3 | 289 | |||
S49491.1 | - | 3 | 359 | |||
Y07503.1 | - | 2 | 568 |

More Information
Other Species Targeted

Gene Ontology Categories:
