siRNA Details
Gene Symbol: Rps7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031570.2 | NP_113758.2 | 6 | 495 | ||
GenBank mRNA | BC060557.1 | - | 6 | 499 | 6888098 | |
BC168722.1 | - | 5 | 479 | 6922220 | ||
FQ210052.1 | - | 6 | 495 | |||
FQ211139.1 | - | 6 | 495 | |||
FQ212121.1 | - | 6 | 495 | |||
FQ217452.1 | - | 6 | 495 | |||
FQ219950.1 | - | 6 | 495 | |||
FQ220886.1 | - | 6 | 543 | |||
FQ221065.1 | - | 6 | 495 | |||
FQ221374.1 | - | 6 | 495 | |||
FQ221761.1 | - | 6 | 495 | |||
FQ221878.1 | - | 6 | 495 | |||
FQ221932.1 | - | 6 | 498 | |||
FQ222169.1 | - | 6 | 495 | |||
FQ222484.1 | - | 6 | 495 | |||
FQ222534.1 | - | 6 | 493 | |||
FQ222918.1 | - | 6 | 495 | |||
FQ222969.1 | - | 3 | 246 | |||
FQ222996.1 | - | 6 | 495 | |||
FQ223010.1 | - | 6 | 495 | |||
FQ223103.1 | - | 6 | 495 | |||
FQ223290.1 | - | 6 | 495 | |||
FQ223392.1 | - | 6 | 495 | |||
FQ223426.1 | - | 6 | 495 | |||
FQ223491.1 | - | 6 | 495 | |||
FQ228541.1 | - | 6 | 511 | |||
X53377.1 | - | 6 | 495 | |||
X56846.1 | - | 5 | 482 |
Gene Symbol: LOC100362830 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729576.4 | XP_002729622.2 | 1 | 495 |

More Information
Other Species Targeted

Gene Ontology Categories:
