siRNA Details
Gene Symbol: Tyms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019179.1 | NP_062052.1 | 2 | 209 | ||
XM_008767420.1 | XP_008765642.1 | 2 | 698 | |||
XM_008767421.1 | XP_008765643.1 | 3 | 254 | |||
GenBank mRNA | BC126093.1 | - | 2 | 223 | 8362547 | |
L12138.1 | - | 2 | 209 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
