siRNA Details
Gene Symbol: Tpsb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019180.2 | NP_062053.2 | 3 | 288 | ||
GenBank mRNA | D38455.1 | - | 4 | 298 | ||
U67909.1 | - | 3 | 289 |

More Information
Other Species Targeted

Gene Ontology Categories:
