siRNA Details
Gene Symbol: Dpp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022850.1 | NP_074041.1 | 22 | 2400 | ||
XM_006235880.2 | XP_006235942.1 | 22 | 6473 | |||
GenBank mRNA | M76426.1 | - | 22 | 2400 | ||
M76427.1 | - | 22 | 2253 |
