siRNA Details
Gene Symbol: Rpl29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017150.1 | NP_058846.1 | 4 | 589 | ||
GenBank mRNA | BC086404.1 | - | 4 | 583 | 7304319 | |
FQ221016.1 | - | 4 | 590 | |||
FQ221647.1 | - | 4 | 596 | |||
FQ222035.1 | - | 4 | 611 | |||
FQ229056.1 | - | 4 | 594 | |||
X68283.1 | - | 4 | 589 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
