siRNA Details
Gene Symbol: Rps14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022672.1 | NP_073163.1 | 2 | 170 | ||
GenBank mRNA | BC058472.1 | - | 1 | 177 | 6918325 | |
FQ210177.1 | - | 2 | 172 | |||
FQ210178.1 | - | 2 | 172 | |||
FQ210421.1 | - | 2 | 172 | |||
FQ210469.1 | - | 2 | 169 | |||
FQ211405.1 | - | 2 | 170 | |||
FQ211449.1 | - | 2 | 170 | |||
FQ211623.1 | - | 2 | 172 | |||
FQ212082.1 | - | 2 | 174 | |||
FQ212093.1 | - | 2 | 170 | |||
FQ212392.1 | - | 2 | 170 | |||
FQ214636.1 | - | 2 | 172 | |||
FQ217229.1 | - | 2 | 170 | |||
FQ217605.1 | - | 2 | 170 | |||
FQ218082.1 | - | 2 | 167 | |||
FQ218760.1 | - | 2 | 172 | |||
FQ219972.1 | - | 2 | 170 | |||
FQ220057.1 | - | 2 | 170 | |||
FQ220332.1 | - | 2 | 172 | |||
FQ220676.1 | - | 2 | 174 | |||
FQ220975.1 | - | 2 | 167 | |||
FQ221057.1 | - | 2 | 170 | |||
FQ221058.1 | - | 2 | 170 | |||
FQ221066.1 | - | 2 | 170 | |||
FQ221125.1 | - | 2 | 172 | |||
FQ221205.1 | - | 2 | 170 | |||
FQ221325.1 | - | 2 | 170 | |||
FQ221335.1 | - | 2 | 170 | |||
FQ221375.1 | - | 2 | 170 | |||
FQ221450.1 | - | 2 | 170 | |||
FQ221468.1 | - | 2 | 170 | |||
FQ221634.1 | - | 2 | 167 | |||
FQ221754.1 | - | 2 | 166 | |||
FQ221835.1 | - | 2 | 172 | |||
FQ221893.1 | - | 2 | 172 | |||
FQ221920.1 | - | 2 | 172 | |||
FQ222066.1 | - | 2 | 167 | |||
FQ222119.1 | - | 2 | 170 | |||
FQ222124.1 | - | 2 | 170 | |||
FQ222159.1 | - | 2 | 172 | |||
FQ222163.1 | - | 2 | 171 | |||
FQ222324.1 | - | 2 | 174 | |||
FQ222370.1 | - | 2 | 170 | |||
FQ222480.1 | - | 2 | 170 | |||
FQ222552.1 | - | 2 | 169 | |||
FQ222574.1 | - | 2 | 170 | |||
FQ222575.1 | - | 1 | 170 | |||
FQ222601.1 | - | 2 | 185 | |||
FQ222667.1 | - | 2 | 170 | |||
FQ222685.1 | - | 2 | 170 | |||
FQ222697.1 | - | 2 | 172 | |||
FQ222714.1 | - | 2 | 170 | |||
FQ222747.1 | - | 2 | 174 | |||
FQ222803.1 | - | 2 | 170 | |||
FQ222840.1 | - | 2 | 173 | |||
FQ222893.1 | - | 2 | 172 | |||
FQ222979.1 | - | 2 | 172 | |||
FQ222986.1 | - | 2 | 167 | |||
FQ223007.1 | - | 2 | 170 | |||
FQ223012.1 | - | 2 | 172 | |||
FQ223069.1 | - | 2 | 172 | |||
FQ223131.1 | - | 2 | 170 | |||
FQ223204.1 | - | 2 | 172 | |||
FQ223283.1 | - | 2 | 170 | |||
FQ223454.1 | - | 2 | 171 | |||
FQ223468.1 | - | 2 | 170 | |||
FQ223480.1 | - | 2 | 170 | |||
FQ223519.1 | - | 2 | 169 | |||
FQ223527.1 | - | 2 | 170 | |||
FQ223570.1 | - | 2 | 170 | |||
FQ224051.1 | - | 2 | 167 | |||
FQ224105.1 | - | 2 | 172 | |||
FQ224302.1 | - | 2 | 171 | |||
FQ224493.1 | - | 2 | 172 | |||
FQ225464.1 | - | 2 | 172 | |||
FQ228360.1 | - | 2 | 155 | |||
FQ228480.1 | - | 2 | 172 | |||
FQ228486.1 | - | 2 | 170 | |||
FQ228559.1 | - | 2 | 174 | |||
FQ228608.1 | - | 2 | 170 | |||
FQ228692.1 | - | 2 | 169 | |||
FQ229006.1 | - | 2 | 176 | |||
FQ229014.1 | - | 2 | 172 | |||
FQ229055.1 | - | 2 | 172 | |||
FQ229061.1 | - | 2 | 172 | |||
FQ229229.1 | - | 2 | 172 | |||
FQ229387.1 | - | 2 | 170 | |||
FQ231306.1 | - | 313 | ||||
X15040.1 | - | 2 | 170 |
Gene Symbol: LOC100911847 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750030.3 | XP_003750078.3 | 1 | 174 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
