siRNA Details
Gene Symbol: Coq3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019187.1 | NP_062060.1 | 1 | 48 | ||
GenBank mRNA | BC081811.1 | - | 213 | |||
BC098666.1 | - | 206 | ||||
BC126094.1 | - | 220 | 8370738 | |||
FQ216025.1 | - | 260 | ||||
L20427.1 | - | 1 | 48 |
