siRNA Details
Gene Symbol: Clec11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012459.1 | NP_001012477.1 | 2 | 321 | ||
GenBank mRNA | AB009246.1 | - | 2 | 203 | ||
BC090344.1 | - | 2 | 321 | 7304238 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
