siRNA Details
Gene Symbol: Stmn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017166.1 | NP_058862.1 | 3 | 231 | ||
XM_006239123.2 | XP_006239185.1 | 3 | 229 | |||
GenBank mRNA | BC062234.1 | - | 3 | 156 | 6917958 | |
FQ211845.1 | - | 3 | 154 | |||
FQ212924.1 | - | 3 | 153 | |||
FQ213053.1 | - | 3 | 154 | |||
FQ214201.1 | - | 3 | 148 | |||
FQ214447.1 | - | 3 | 154 | |||
FQ220047.1 | - | 3 | 154 | |||
J04979.1 | - | 3 | 231 | |||
M27876.1 | - | 2 | 174 | |||
X94914.1 | - | 1 | 63 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_019641, AK002606, BC031831, AK014108, X94915, and others.
Gene Ontology Categories:
