siRNA Details
Gene Symbol: Apcs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017170.2 | NP_058866.2 | 2 | 284 | ||
XM_008769786.1 | XP_008768008.1 | 3 | 593 | |||
GenBank mRNA | M83177.1 | - | 2 | 284 | ||
X55761.1 | - | 2 | 366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
