siRNA Details
Gene Symbol: Prkce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017171.1 | NP_058867.1 | 9 | 1485 | ||
GenBank mRNA | AY642593.1 | - | 9 | 1299 | ||
M15523.1 | - | 2 | 104 | |||
M18331.1 | - | 9 | 1485 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
