siRNA Details
Gene Symbol: Pkn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017175.2 | NP_058871.2 | 4 | 695 | ||
XM_006255245.2 | XP_006255307.1 | 4 | 1735 | |||
GenBank mRNA | BC061836.1 | - | 4 | 596 | 5599989 | |
D26180.1 | - | 4 | 646 | |||
FQ222217.1 | - | 4 | 695 | |||
L35634.1 | - | 4 | 538 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
