siRNA Details
Gene Symbol: Smad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277450.1 | NP_001264379.1 | 10 | 1430 | ||
NM_019191.2 | NP_062064.1 | 10 | 1458 | |||
XM_006254945.2 | XP_006255007.1 | 9 | 1454 | |||
XM_006254946.2 | XP_006255008.1 | 10 | 1171 | |||
GenBank mRNA | AB010147.1 | - | 9 | 1232 | ||
AB017912.1 | - | 10 | 1324 | |||
AF056001.1 | - | 10 | 1358 | |||
BC127497.1 | - | 10 | 1302 | 8364718 |
