siRNA Details
Gene Symbol: Bmp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017178.1 | NP_058874.1 | 3 | 1179 | ||
XM_008762246.1 | XP_008760468.1 | 2 | 2448 | |||
GenBank mRNA | L20678.1 | - | 3 | 1179 | ||
Z25868.1 | - | 3 | 1179 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
