siRNA Details
Gene Symbol: Cxcr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017183.1 | NP_058879.1 | 2 | 258 | ||
XM_008767213.1 | XP_008765435.1 | 2 | 584 | |||
XM_008767214.1 | XP_008765436.1 | 2 | 1103 | |||
XM_008767215.1 | XP_008765437.1 | 2 | 424 | |||
GenBank mRNA | X77797.1 | - | 2 | 258 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
