siRNA Details
Gene Symbol: Pla2g2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019202.2 | NP_062075.1 | 6 | 1700 | ||
XM_006239142.2 | XP_006239204.1 | 6 | 1718 | |||
XM_006239143.2 | XP_006239205.1 | 5 | 1479 | |||
XM_008764212.1 | XP_008762434.1 | 6 | 1793 | |||
XM_008764213.1 | XP_008762435.1 | 6 | 1571 | |||
XM_008764214.1 | XP_008762436.1 | 6 | 3242 | |||
GenBank mRNA | BC097325.1 | - | 6 | 1700 | 7456563 | |
U07798.1 | - | 6 | 2028 |
