siRNA Details
Gene Symbol: Pla2g2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019202.2 | NP_062075.1 | 5 | 658 | ||
XM_006239142.2 | XP_006239204.1 | 5 | 676 | |||
XM_006239143.2 | XP_006239205.1 | 4 | 437 | |||
XM_008764212.1 | XP_008762434.1 | 5 | 751 | |||
XM_008764213.1 | XP_008762435.1 | 5 | 529 | |||
XM_008764214.1 | XP_008762436.1 | 5 | 2200 | |||
GenBank mRNA | BC097325.1 | - | 5 | 658 | 7456563 | |
U07798.1 | - | 5 | 985 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
