siRNA Details
Gene Symbol: Tnni2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017185.1 | NP_058881.1 | 5 | 250 | ||
GenBank mRNA | FQ216337.1 | - | 5 | 292 | ||
FQ217026.1 | - | 6 | 292 | |||
FQ217309.1 | - | 5 | 283 | |||
FQ217454.1 | - | 6 | 300 | |||
FQ217985.1 | - | 6 | 292 | |||
FQ218058.1 | - | 5 | 324 | |||
FQ223581.1 | - | 6 | 292 | |||
FQ223772.1 | - | 6 | 298 | |||
FQ224443.1 | - | 6 | 293 | |||
FQ224464.1 | - | 6 | 292 | |||
FQ224489.1 | - | 6 | 292 | |||
M73701.1 | - | 5 | 250 |
