siRNA Details
Gene Symbol: Col1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053304.1 | NP_445756.1 | 49, 50 | 4091 | ||
GenBank mRNA | BC133728.1 | - | 49, 50 | 4075 | 7125649 | |
KJ696743.1 | - | 5, 6 | 687 | |||
Z78279.1 | - | 49, 50 | 3980 |
